Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

90220-001

Sigma-Aldrich

RNAstable®

greener alternative

96-well plate, for room temp preservation of RNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

grade

for molecular biology

form

dry

usage

96 reactions

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

greener alternative category

storage temp.

room temp

General description

SampleMatrix® by Biomatrica® is a proprietary core technology to stabilize and protect biological materials at room temperature without degradation. The SampleMatrix technology was designed by combining extremophile biology that enables long-term survival of organisms in extremely dry environments, with synthetic chemistry.

RNAstable® preserves RNA samples at room temperature. Each tube or plate contains RNAstable® as a coating at the bottom of the tube or well, which protects picogram to microgram amounts of RNA. This medium is completely dissolvable and ensures total and quantitative sample recovery.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product was Designed for Energy Efficiency. To see how Stanford incorporated Biomatrica Technology and saved substantial amounts of energy, see their published study:

Biomatrica technology enables safe storage of biological material at room temperature.

Features and Benefits

  • Preserves RNA at ambient temperatures without degradation
  • Eliminates the need to store RNA in freezers or to ship on dry ice
  • Concentrates RNA more effectively than other methods
  • Compatible with most downstream applications

Packaging

  • (1) RNAstable® 96-Well Plate
  • Hinged lid provides additional sample protection
  • (1) tube of sterile water
  • Quick Reference Protocol
  • Barcode-labeled for sample tracking
  • Enclosed in re-sealable moisture-barrier foil pouch

Legal Information

Biomatrica is a registered trademark of Biomatrica
RNAstable is a registered trademark of Biomatrica
SampleMatrix is a registered trademark of Biomatrica

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen K Gire et al.
Science (New York, N.Y.), 345(6202), 1369-1372 (2014-09-13)
In its largest outbreak, Ebola virus disease is spreading through Guinea, Liberia, Sierra Leone, and Nigeria. We sequenced 99 Ebola virus genomes from 78 patients in Sierra Leone to ~2000× coverage. We observed a rapid accumulation of interhost and intrahost
Stefano Begolo et al.
Lab on a chip, 13(22), 4331-4342 (2013-09-24)
This paper describes a microfluidic device for dry preservation of biological specimens at room temperature that incorporates chemical stabilization matrices. Long-term stabilization of samples is crucial for remote medical analysis, biosurveillance, and archiving, but the current paradigm for transporting remotely
Cornelia Blaser et al.
PloS one, 6(3), e17840-e17840 (2011-03-18)
Streptococcus pneumoniae is the most common pathogen causing non-epidemic bacterial meningitis worldwide. The immune response and inflammatory processes contribute to the pathophysiology. Hence, the anti-inflammatory dexamethasone is advocated as adjuvant treatment although its clinical efficacy remains a question at issue.
Gilberto E Hernandez et al.
BioTechniques, 47(2), 667-668 (2009-09-10)
RNA integrity is a critical factor in obtaining meaningful gene expression data. Current methodologies rely on maintaining samples in cold environments during collection, transport, processing, and storage procedures, which are also extremely time-sensitive. Several RNA storage products are commercially available
Wen Huang et al.
PloS one, 7(1), e30244-e30244 (2012-02-01)
In this study, mRNA-Seq was used to characterize and compare the leukocyte transcriptomes from two taurine breeds (Holstein and Jersey), and one indicine breed (Cholistani). At the genomic level, we identified breed-specific base changes in protein coding regions. Among 7,793,425

Articles

RNAstable® preserves RNA via anhydrobiosis principles, enabling ambient temp storage and shipping.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service