Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89101

Sigma-Aldrich

Atto Rho11 NHS ester

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

product line

BioReagent

Quality Level

form

solid

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 571 nm; λem 595 nm

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto Rho11 is a new rhodamine dye featuring a functionality for coupling to biomolecules such as DNA, RNA or proteins. The label shows very high fluorescence efficiency and exceptionally high photostability.
After coupling to a substrate Atto Rho11 carries a net electrical charge of +1.

The dye is moderately hydrophilic.find more information here

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service