Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88886

Sigma-Aldrich

9-(Bromomethyl)acridine

BioReagent, suitable for fluorescence, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10BrN
CAS Number:
Molecular Weight:
272.14
Beilstein:
159526
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥97.0% (HPLC)

form

powder

mp

160-165 °C

fluorescence

λex 335 nm; λem 469 nm in 0.1 M phosphate pH 7.0 (after derivatization with glutathione)

suitability

suitable for fluorescence

SMILES string

BrCc1c2ccccc2nc3ccccc13

InChI

1S/C14H10BrN/c15-9-12-10-5-1-3-7-13(10)16-14-8-4-2-6-11(12)14/h1-8H,9H2

InChI key

MZFYKBHQWLWIBI-UHFFFAOYSA-N

Analysis Note

In addition to the emission maximum at 447 nm, there are lower maxima at 468 and 413 nm.

Other Notes

HPLC derivatizing agent for fluorescent labeling of carboxylic acids, e.g. fatty acids

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K. Akasaka et al.
Analytical Letters, 20, 1581-1581 (1987)
Wakako Tsuzuki et al.
The Analyst, 127(5), 669-673 (2002-06-26)
For the purpose of deducing the digestive behavior of dietary fat in the digestive organs, a fluorimetric method for the measurement of hydrolysis by porcine pancreatic lipase was performed using intermediate- and long- acyl chain glycerides as substrates. Insoluble glycerides
F A van der Horst et al.
Journal of chromatography, 500, 443-452 (1990-02-02)
The on-line use of micellar phase-transfer catalysis is described for the automated reversed-phase high-performance liquid chromatographic (RP-HPLC) determination of free fatty acids in plasma; minimum manual sample handling is involved. After diluting plasma ten-fold with the aqueous micellar system, which
Yi-Shan Li et al.
Talanta, 218, 121143-121143 (2020-08-17)
Dipeptidyl peptidase 4 (DPP-4) inhibitors are incretin-based medications used as oral antidiabetic agents for the treatment of type 2 diabetes. However, DPP-4 inhibitors produce side effects like acute pancreatitis, upper respiratory tract infection, nasopharyngitis, urinary tract infection, serious allergies, cardiovascular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service