Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88815

Sigma-Aldrich

22,23-Dihydroergocalciferol

≥97.0% (HPLC)

Synonym(s):

(3β,5Z,7E)-9,10-Secoergosta-5,7,10(19)-trien-3-ol, 22,23-Dihydrovitamin D2, 22-Dihydroergocalciferol, Vitamin D4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H46O
CAS Number:
Molecular Weight:
398.66
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.79

Assay

≥97.0% (HPLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CC(C)[C@@H](C)CC[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C3\C[C@@H](O)CCC3=C

InChI

1S/C28H46O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h12-13,19-20,22,25-27,29H,4,7-11,14-18H2,1-3,5-6H3/b23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1

InChI key

DIPPFEXMRDPFBK-JPWDPSJFSA-N

General description

22,23-Dihydroergocalciferol, also known as vitamin D4, is derived from fungi such as yeast. It is synthesized from the precursor ergosta-5,7-dienol (22,23-dihydroergosterol).[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin D4 in mushrooms
Phillips KM, et al.
PLoS ONE, 7(8), e40702-e40702 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service