Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83392

Sigma-Aldrich

Atto-Dino 4 solution

BioReagent, suitable for fluorescence, in DMSO

Synonym(s):

Nucleic acid stain 6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500

product line

BioReagent

form

solution

manufacturer/tradename

ATTO-TEC GmbH

concentration

in DMSO

solubility

DMSO: soluble

suitability

suitable for fluorescence

storage temp.

−20°C

Physical form

0.5% solids in aquaeous buffer, pH 7.0

Analysis Note

Atto-Dino 4 is a cell permeable homodimer. In aqeous solution it does not show any fluorescence. In presence of DNA it shows a bright fluorescence with quantum yield above 70%. Maximum of absorption lies at 485 nm, exctinction coefficient 140000 M-1cm-1. Maximum of emission shifts from 523 nm to 572 nm if bound to DNA.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service