Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83263

Sigma-Aldrich

BICINE buffer Solution

BioUltra, for molecular biology, 1 M in H2O

Synonym(s):

N,N-Bis(2-hydroxyethyl)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO4
CAS Number:
Molecular Weight:
163.17
Beilstein:
1769362
MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
PubChem Substance ID:

grade

for molecular biology

product line

BioUltra

concentration

1 M in H2O

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

pH

5.0±0.2

density

1.05 g/mL at 20 °C

λ

neat

UV absorption

λ: 260 nm Amax: <0.07
λ: 280 nm Amax: <0.04

suitability

in accordance for filter test

SMILES string

OCCN(CCO)CC(O)=O

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ito et al.
Clinica chimica acta; international journal of clinical chemistry, 115(2), 135-144 (1981-09-10)
Serum guanase activity has been considered as a possible specific indicator of hepatocellular diseases. However, no suitable method is available for routine clinical determination of serum guanase activity. Conventional assay methods are troublesome and inaccurate, since guanine and 8-azaguanine, the
Yoshiteru Seo et al.
Magnetic resonance in medicine, 65(4), 1005-1012 (2011-03-18)
The toxicity of free Mn(2+) is a bottleneck for the in vivo application of manganese ion enhanced MRI. To reduce free Mn(2+) concentration ([Mn(2+) ]), a low affinity chelate reagent: N,N-bis(2-hydroxyethyl)glycine (bicine) was used. Considering the conditional association constant of
The myosin dimer: an intermediate in the self-assembly of the thick filament of vertebrate skeletal muscle.
J S Davis et al.
FEBS letters, 140(2), 293-297 (1982-04-19)
R N Roy et al.
Cryobiology, 21(6), 672-681 (1984-12-01)
The establishment of the pH (designated pH*) of a standard buffer solution suitable as a pH reference in 30, 40, and 50 mass% dimethyl sulfoxide (DMSO)/H2O mixtures at temperatures in the range -20 to 0 degrees C is reported. The
S Ito et al.
The Histochemical journal, 16(5), 489-499 (1984-05-01)
Histochemical studies of human guanase (guanine deaminase) have seldom been undertaken, in part because of technical difficulties which result in heavy background staining. In this report, we describe a modified procedure in which the methodological inadequacies have been overcome. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service