Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82485

Sigma-Aldrich

Protein A (extracellular) from Staphylococcus aureus

lyophilized, ≥90% protein basis

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

unconjugated

Assay

≥90% protein basis

form

fibers

quality

lyophilized

mol wt

Mr ~41000

species reactivity

pig, cat, monkey, guinea pig, rabbit, dog, mouse, human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

Staphylococcus aureus subsp. aureus NCTC 8325 ... SAOUHSC_00069(3919448)

Looking for similar products? Visit Product Comparison Guide

General description

Protein A is a highly stable cell surface receptor produced by several strains of Staphylococcus aureus. This protein is capable of binding to the Fc portion of immunoglobulins, especially IgGs, from a large number of species. One protein A molecule has been shown to bind at least 2 molecules of IgG simultaneously. The IgG binding domain of Protein A consists of three anti-parallel alpha-helicies, the third of which is disrupted when the protein is complexed with the Fc region of the immunoglobulins. Protein A will bind the Fc portion of human IgG subclasses, IgM, IgA and IgE; and mouse IgG1 (weakly), IgG2a, and IgG2b. Protein A also associates with IgGs from other species, including monkey, rabbit, pig, guinea pig, dog, and cat.
Protein A is involved in regulating anti-tumor, toxic, and carcinogenic functions. In addition to acting as an immunomodulator, it also has antifungal and antiparasitic properties.

Application

Protein A may be conjugated with various reporter molecules such as fluorescent dyes (FITC), enzyme markers (peroxidase, beta-galactosidase, alkaline phosphatase), biotin, and colloidal gold without affecting the antibody binding site on the molecule. These conjugates may be used to detect immunoglobulins using immunochemical assays such as western blotting, immunohistochemistry, and ELISA. In addition, protein A may also be immobilized on a solid supports like agarose or acrylic beads for the purification of polyclonal or monoclonal immunoglobulins. It is also routinely used for immunoprecipitation assays. Furthermore, protein A has been used for affinity chromatography and SDS-PAGE assays[1].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Protein A: nature′s universal anti-antibody - a review on properties and applications of protein A[2][3]

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Surolia et al.
Trends in Biochemical Sciences, 7, 74-74 (1982)
J I Flock et al.
The EMBO journal, 6(8), 2351-2357 (1987-08-01)
The gene encoding the fibronectin-binding protein (FNBP) from Staphylococcus aureus strain 8325-4 was isolated from a gene bank in pBR322. The original clone, containing a 6.5-kb insert, gave a functional product present in the periplasm of Escherichia coli. Analysis of
R Lindmark et al.
Journal of immunological methods, 62(1), 1-13 (1983-08-12)
The use of protein A from S. aureus (SpA) as an anti-IgG reagent in immunological techniques has extended in recent years, together with knowledge about its interaction with immunoglobulins of different species. Current data with respect to the binding of
Romel Rosales Ramirez et al.
Journal of virology, 93(4) (2018-11-23)
Dengue virus (DENV) is a mosquito-borne virus of the family Flaviviridae The RNA viral genome encodes three structural and seven nonstructural proteins. Nonstructural protein 1 (NS1) is a multifunctional protein actively secreted in vertebrate and mosquito cells during infection. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service