Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81227

Sigma-Aldrich

Poly(ethylene glycol)

BioUltra, 3,000

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$72.80
1 KG
$216.00
2.5 KG
$527.00

$72.80


Available to ship onApril 10, 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
$72.80
1 KG
$216.00
2.5 KG
$527.00

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.26

$72.80


Available to ship onApril 10, 2025Details


Request a Bulk Order

product line

BioUltra

form

solid

mol wt

Mr 2700-3300

impurities

insoluble matter, passes filter test
≤0.001% peroxides (as H2O2)

pH

5.5-7.0 (25 °C, 50 mg/mL in H2O)

solubility

H2O: 50 mg/mL at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

50 mg/mL in H2O

UV absorption

λ: 260 nm Amax: 0.09
λ: 280 nm Amax: 0.03

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a non-ionic hydrophilic polymer and is available in different molecular weights. It helps in the purification and crystal growth of proteins and nucleic acids. PEG and dextran together result in aqueous polymer two phase system, which is required for the purification of biological materials. PEG also interacts with cell membrane, thereby allowing cell fusion.[1][2]

Application

Poly(ethylene glycol) (PEG) has been used to study PEG-β lactoglobulin interaction.[3] It has also been used to study the interaction of serum albumins with PEG.[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microscopic and thermodynamic analysis of PEG??-lactoglobulin interaction.
Bekale L, et al.
Royal Society of Chemistry Advances, 4, 31084-31093 (2014)
Poly(Ethylene Glycol) Chemistry: Biotechnical and Biomedical Applications (2013)
Poly (ethylene glycol) in drug delivery: pros and cons as well as potential alternatives.
Katrin K, et al.
Angewandte Chemie (International Edition in English), 49, 6288-6308 (2010)
L Bekale et al.
Colloids and surfaces. B, Biointerfaces, 130, 141-148 (2015-04-14)
We investigated the interaction between polyethylene (glycol) (PEG) and human (HSA) and bovine serum albumin (BSA) in aqueous solution, using multiple spectroscopic methods and molecular modeling. The two important polymer characteristics, size and PEG hydrophobic end-group are studied in order
Thomas McNerney et al.
mAbs, 7(2), 413-428 (2015-02-24)
High titer (>10 g/L) monoclonal antibody (mAb) cell culture processes are typically achieved by maintaining high viable cell densities over longer culture durations. A corresponding increase in the solids and sub-micron cellular debris particle levels are also observed. This higher burden

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service