Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80999

Sigma-Aldrich

1L-epi-2-Inosose

≥98.0% (HPLC)

Synonym(s):

(2R,3S,4S,5S,6S)-2,3,4,5,6-Pentahydroxycyclohexanone, 2L-2,3,4,6/5-Pentahydroxycyclohexanone, D-epi-4-Inosose, L-epi-2-Inosose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14
Beilstein:
3199555
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

Quality Level

Assay

≥98.0% (HPLC)

form

crystals

optical activity

[α]/D -4.3±1.3°, c = 1 in H2O

storage temp.

2-8°C

InChI

1S/C6H10O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-5,7-11H/t1-,2-,3-,4+,5-/m0/s1

InChI key

VYEGBDHSGHXOGT-PKTMPMCPSA-N

Biochem/physiol Actions

Metabolite of inositol metabolic pathways

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics and mechanism of the chromic oxidation of myo-inositol
Santoro, M., et al.
Polyhedron, 26, 169-177 (2007)
(-)-epi-Inosose-2
Acta Crystallographica Section C, Structural Chemistry, C56(12), e584-e585 (2000)
Bacterial enzyme preparations oxidizing inositol and their inhibition by colchicine.
R E FRANZL et al.
Nature, 168(4283), 955-957 (1951-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service