Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80841

Sigma-Aldrich

Lipase, immobilized on Immobead 150 from Candida rugosa

≥70 U/g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

powder

specific activity

≥70 U/g

storage temp.

2-8°C

Application

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Biochem/physiol Actions

Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol butyric acid per minute at pH 7.5 and 40°C (tributyrin, Cat. No. 91010, as substrate)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Timper et al.
Cell metabolism, 31(6), 1189-1205 (2020-05-21)
Astrocytes represent central regulators of brain glucose metabolism and neuronal function. They have recently been shown to adapt their function in response to alterations in nutritional state through responding to the energy state-sensing hormones leptin and insulin. Here, we demonstrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service