Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79273

Sigma-Aldrich

Kaempferol 3-O-β-D-glucuronide

≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O12
CAS Number:
Molecular Weight:
462.36
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥97.0% (HPLC)

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)cc4

InChI

1S/C21H18O12/c22-8-3-1-7(2-4-8)17-18(13(25)12-10(24)5-9(23)6-11(12)31-17)32-21-16(28)14(26)15(27)19(33-21)20(29)30/h1-6,14-16,19,21-24,26-28H,(H,29,30)/t14-,15-,16+,19-,21+/m0/s1

InChI key

FNTJVYCFNVUBOL-ZUGPOPFOSA-N

Biochem/physiol Actions

Flavonoid glycoside found in vegetables.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of a quantitative HPLC determination of potentially anticarcinogenic flavonoids in vegetables and fruits.
Hertog, M.G.L., et al.
Journal of Agricultural and Food Chemistry, 40, 1591-1598 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service