Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77958

Sigma-Aldrich

Abberior®CAGE 590, NHS-ester

ideal for single-molecule siwtching microscopy (e.g. PALM, STORM, GSDIM)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

form

solid

solubility

DMF: 0.25 mg/mL, clear

fluorescence

λex 590 nm; λem 600-620 nm in PBS, pH 7

storage temp.

−20°C

General description

Abberior CAGE 590 is a masked dye which is initially colorless and nonfluorescent. When illuminated with UV light it undergoes a rapid uncaging reaction and releases a highly fluorescent dye. The dye opens up the red fluorescent regime for CAGE dyes. It can most effectively be excited from 560 to 600nm.

Abberior CAGE 590 is designed for single-molecule photoswitching microscopy modes such as PALM, STORM and GSDIM and, in its uncaged form, also performs well in STED microscopy. Further, after photoactivation, Abberior CAGE markers can be tracked in confocal and epifluorescence microscopy to analyse molecular dynamics such as diffusion, flow directions and velocities, e.g. are tracking experiments in which the fluorescence signal is observed over time after the initial release of the fluorophore.

Key Features
  • First CAGE dye in the red fluorescent range
  • Ideal for PALM, STORM, GSDIM
  • Well suited as 2nd STED color (after uncaging)


Absorption Maximum, λmax: 262, 325, 351 nm (non-activated, PBS, pH 7.4)
595 nm (photoactivated, PBS, pH 7.4)
Extinction Coefficient, ε(λmax): 75,000 M-1cm-1 (photoactivated, PBS, pH 7.4)
Fluorescence Maximum, λfl: 615 nm (photoactivated, PBS, pH 7.4)
Photoactivation Wavelength:360 - 440 nm
Recommended STED Wavelength, λSTED: 685 −715 nm
Fluorescence Quantum Yield, η: 0.80 (after photoactivation, PBS, pH 7.47)

Application

Abberior® CAGE 590 anti-rabbit antibody has been used as a secondary antibody for spectral-FPALM (fluorescence photo-activation localization microscopy) imaging of NIH-3T3 cells.[1]

Suitability

Designed and tested for fluorescent super-resolution microscopy

Analysis Note

degree of coupling: report results

Other Notes

For more information on Abberior® Dyes, please visit our overview.

Legal Information

abberior is a registered trademark of Abberior GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Mlodzianoski et al.
PloS one, 11(3), e0147506-e0147506 (2016-03-24)
Localization microscopy can image nanoscale cellular details. To address biological questions, the ability to distinguish multiple molecular species simultaneously is invaluable. Here, we present a new version of fluorescence photoactivation localization microscopy (FPALM) which detects the emission spectrum of each

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service