Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77404

Sigma-Aldrich

2′,3′-Dideoxyadenosine 5′-triphosphate trilithium salt

≥95.0% (HPLC)

Synonym(s):

ddATP-Li3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13Li3N5O11P3
CAS Number:
Molecular Weight:
492.98
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥95.0% (HPLC)

form

powder

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Li+].[Li+].[Li+].Nc1ncnc2n(cnc12)[C@H]3CC[C@@H](COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)O3

InChI

1S/C10H16N5O11P3.3Li/c11-9-8-10(13-4-12-9)15(5-14-8)7-2-1-6(24-7)3-23-28(19,20)26-29(21,22)25-27(16,17)18;;;/h4-7H,1-3H2,(H,19,20)(H,21,22)(H2,11,12,13)(H2,16,17,18);;;/q;3*+1/p-3/t6-,7+;;;/m0.../s1

InChI key

ZTVIJXZPFNMGOR-PXJNTPRPSA-K

Looking for similar products? Visit Product Comparison Guide

Physical form

aqueous solution (~10 mM)

comparable product

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service