Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77371

Sigma-Aldrich

Protein Quantification Kit, wide range

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for 500 tests

shelf life

limited shelf life, expiry date on the label

shipped in

dry ice

storage temp.

2-8°C

General description

This kit is a complete, ready-to-use microtiter plate assay kit for the colorimetric detection of proteins. It is based on the reducing reaction of a tetrazolium salt under basic conditions. Tetrazolium salt is easily reduced with sugar residues of proteins and generates formazan dye. The maximum wavelength of the WST-8 formazan dye is 650 nm at pH 12.5.
The detection range of this protein quantification kit is from 50 μg/ml to 5000 μg/ml (incubation time 30 min).

Components

Contents: WST-8 Solution 10 ml, Buffer Solution 100 ml, BSA Standard Solution (10 mg/ml) 1.5 mL (500 tests)

Kit Components Only

Product No.
Description

  • WST-8 Solution 10 mL

  • Buffer Solution 100 mL

  • BSA Standard Solution (10 mg/ml) 1.5 mL

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dario Lombardi et al.
Analytical and bioanalytical chemistry, 399(1), 347-352 (2010-11-05)
In this study, we give the proof of concept for a method to determine binding constants of compounds in solution. By implementing a technique based on magnetic beads with a microfluidic device for segmented flow generation, we demonstrate, for individual

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service