Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76667

Sigma-Aldrich

D-myo-Inositol 1,2,3,5,6-pentakis-phosphate decasodium salt

≥80% (HPLC)

Synonym(s):

L-myo-Inositol 1,2,3,4,5-pentakis-phosphate decasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7Na10O21P5
Molecular Weight:
799.87
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥80% (HPLC)

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].O[C@H]1[C@H](OP([O-])([O-])=O)[C@@H](OP([O-])([O-])=O)[C@H](OP([O-])([O-])=O)[C@@H](OP([O-])([O-])=O)[C@H]1OP([O-])([O-])=O

InChI

1S/C6H17O21P5.10Na/c7-1-2(23-28(8,9)10)4(25-30(14,15)16)6(27-32(20,21)22)5(26-31(17,18)19)3(1)24-29(11,12)13;;;;;;;;;;/h1-7H,(H2,8,9,10)(H2,11,12,13)(H2,14,15,16)(H2,17,18,19)(H2,20,21,22);;;;;;;;;;/q;10*+1/p-10/t1-,2-,3-,4+,5-,6-;;;;;;;;;;/m0........../s1

InChI key

GFGCWCIISVRGOQ-HSYGTAAMSA-D

Biochem/physiol Actions

Intermediate in inositol phosphate metabolism. Inositides have an important impact on diverse areas of cellular regulation.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher J Barker et al.
Methods in molecular biology (Clifton, N.J.), 645, 1-19 (2010-07-21)
Inositides have an important impact on diverse areas of cellular regulation. However, since this area has grown exponentially from the mid 1980s onwards, many workers find themselves relatively new to the field. In this chapter, we establish a broad foundation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service