Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75925

Sigma-Aldrich

Oxonol V

for fluorescence, ≥95% (UV)

Synonym(s):

1,5-Penta-1,3-dien-1-yl-5-ylidene-bis(5-oxo-3-phenyl-4-isoxazole), 4-[5-(5-Hydroxy-3-phenyl-4-isoxazolyl)-2,4-pentadienylidene]-3-phenyl-5(4H)-isoxazolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H16N2O4
CAS Number:
Molecular Weight:
384.38
Beilstein:
575970
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

for fluorescence

Assay

≥95% (UV)

suitability

in accordance for fluorescence
suitable for fluorescence

shipped in

wet ice

storage temp.

−20°C

SMILES string

Oc1onc(-c2ccccc2)c1\C=C\C=C\C=C3/C(=O)ON=C3c4ccccc4

InChI

1S/C23H16N2O4/c26-22-18(20(24-28-22)16-10-4-1-5-11-16)14-8-3-9-15-19-21(25-29-23(19)27)17-12-6-2-7-13-17/h1-15,26H/b9-3+,14-8+,19-15-

InChI key

NEDZCQDBZLHEGJ-UFRBDIPQSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Potential sensitive probe. Plasma membrane potential of neutrophils generated by the Na+ pump; Spectral properties

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C L Bashford et al.
Biochimica et biophysica acta, 817(1), 174-180 (1985-07-11)
The plasma membrane potential of human neutrophils was monitored using the anionic dye oxonol-V. The cells maintain a potential of -75 +/- 17 mV when suspended in physiological saline solutions. The cells are scarcely depolarized by extracellular K+ and the
J C Smith et al.
Biochemistry, 15(23), 5094-5105 (1976-11-16)
The direct synthesis of bis]3-phenyl-5-oxoisoxazol-4-yl]pentamethineoxonol, which is shown to be the fluorescent probe OX-V (formerly MC-V), is described. The emission lifetime (0.9 +/- 0.1 ns) and the spectral properties of this dye in a number of systems are presented as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service