Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75824

Sigma-Aldrich

Tracy 652 Protein Labeling Kit

suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

fluorescence

λex 648 nm; λem 670 nm in PBS (0.1 M)

suitability

suitable for fluorescence

storage temp.

2-8°C

Components

This kit contains sufficient amounts of reactive dye, buffers, and protein purification sets for performing five labeling reactions (1 mg protein each) and for the subsequent purification of the labeled protein.

Other Notes

Tracy 652 is a NIR fluorescent dye that is highly water-soluble and stable. The spectral range above 650 nm is of particular interest in biology analysis and imaging due to the reduced background and improved sensitivity.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service