Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75081

Sigma-Aldrich

Octyl D-glucopyranoside

mixture of anomers, α-anomer/β-anomer ~7:3, ~80% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28O6
CAS Number:
Molecular Weight:
292.37
Beilstein:
4864266
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

description

non-ionic

Assay

~80% (TLC)

form

paste

mol wt

292.37 g/mol

loss

≤10% loss on drying

color

beige

SMILES string

CCCCCCCCOC1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14?/m1/s1

InChI key

HEGSGKPQLMEBJL-RQICVUQASA-N

Other Notes

For the crystallisation of membrane proteins[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriela Camporeale et al.
Redox biology, 11, 38-50 (2016-11-20)
Infection with oncogenic human papillomavirus induces deregulation of cellular redox homeostasis. Virus replication and papillomavirus-induced cell transformation require persistent expression of viral oncoproteins E7 and E6 that must retain their functionality in a persistent oxidative environment. Here, we dissected the
Kristian Parey et al.
Science advances, 5(12), eaax9484-eaax9484 (2019-12-18)
Respiratory complex I is a redox-driven proton pump, accounting for a large part of the electrochemical gradient that powers mitochondrial adenosine triphosphate synthesis. Complex I dysfunction is associated with severe human diseases. Assembly of the one-megadalton complex I in the
Joan E Nichols et al.
Virology, 534, 80-86 (2019-06-21)
The immunopathological mechanisms as well as the role played by influenza A virus infection of human leukocytes and induction of apoptosis have not been fully elucidated. We confirm here that the percentage of cells that are infected is less than
Ana Sofia Brito et al.
PLoS genetics, 16(2), e1008634-e1008634 (2020-02-19)
The dimorphic transition from the yeast to the filamentous form of growth allows cells to explore their environment for more suitable niches and is often crucial for the virulence of pathogenic fungi. In contrast to their Mep1/3 paralogues, fungal Mep2-type
Zhao Zhang et al.
Nature chemistry, 9(7), 653-659 (2017-06-24)
The diverse structure and regulated deformation of lipid bilayer membranes are among a cell's most fascinating features. Artificial membrane-bound vesicles, known as liposomes, are versatile tools for modelling biological membranes and delivering foreign objects to cells. To fully mimic the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service