Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74758

Sigma-Aldrich

8-Octadecyloxypyrene-1,3,6-trisulfonic acid trisodium salt

suitable for fluorescence, ≥98.0% (TLC)

Synonym(s):

PTS18, Trisodium 8-octadecyloxypyrene-1,3,6-trisulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H43Na3O10S3
CAS Number:
Molecular Weight:
776.86
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98.0% (TLC)

mp

265 °C (dec.)
265 °C (lit.)

solubility

DMF: soluble
DMSO: soluble
H2O: soluble
methanol: soluble

fluorescence

λex 404 nm; λem 434 nm in H2O

suitability

suitable for fluorescence

SMILES string

[Na+].[Na+].[Na+].CCCCCCCCCCCCCCCCCCOc1cc(c2ccc3c(cc(c4ccc1c2c34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C34H46O10S3.3Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22-44-29-23-30(45(35,36)37)26-20-21-28-32(47(41,42)43)24-31(46(38,39)40)27-19-18-25(29)33(26)34(27)28;;;/h18-21,23-24H,2-17,22H2,1H3,(H,35,36,37)(H,38,39,40)(H,41,42,43);;;/q;3*+1/p-3

InChI key

IABMEQMYXMSMGI-UHFFFAOYSA-K

Other Notes

Highly water soluble superpolar fluorescent lipid probe for the sensitive fluorimetric assay of cationic surfactants

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Marhold et al.
Fresenius Journal of Analytical Chemistry, 336, 111-111 (1990)

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service