Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74436

Sigma-Aldrich

Nonyl-β-D-1-thiomaltoside

≥98% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H40O10S
CAS Number:
Molecular Weight:
484.60
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Assay

≥98% (GC)

form

powder

optical activity

[α]20/D +36±1°, c = 0.1% in methanol

mol wt

484.60 g/mol

CMC

3.2

storage temp.

2-8°C

SMILES string

CCCCCCCCCS[C@@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O

InChI

1S/C21H40O10S/c1-2-3-4-5-6-7-8-9-32-21-18(28)16(26)19(13(11-23)30-21)31-20-17(27)15(25)14(24)12(10-22)29-20/h12-28H,2-11H2,1H3/t12-,13-,14-,15+,16-,17-,18-,19-,20-,21+/m1/s1

InChI key

AFOMEWLJBOWKCP-OHQXVXRSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

n-Nonyl-β-D-thiomaltoside is a newly developed detergent whose lipophilic moiety is maltose. This reagent is suitable for isolating proteins inside of cell membranes. The activity of protein isolated using n-Nonyl-β-D-thiomaltoside is higher than those isolated using n-Octyl-β-D-thioglucoside or n-Heptyl-β-D-thioglucoside. As the CMC value of this detergent is 2.4 mM, it is easy to remove from protein solutions by dialysis.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service