Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73156

Sigma-Aldrich

N-Methyl-L-serine

≥98% (TLC)

Synonym(s):

(S)-3-Hydroxy-2-(methylamino)propionic acid, N-Me-Ser-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO3
CAS Number:
Molecular Weight:
119.12
Beilstein:
2498641
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (TLC)

form

powder

optical activity

[α]/D +9.5±1.0°, c = 1 in 1 M HCl

composition

carbon, 39.5-41.1%
nitrogen, 11.5-12.0%

color

white

mp

204 °C

SMILES string

CN[C@@H](CO)C(O)=O

InChI

1S/C4H9NO3/c1-5-3(2-6)4(7)8/h3,5-6H,2H2,1H3,(H,7,8)/t3-/m0/s1

InChI key

PSFABYLDRXJYID-VKHMYHEASA-N

Biochem/physiol Actions

N-Methyl amino acids play an important role in investigations on prevention or therapy of Alzheimer′s disease[1], neurofibrillary tangles or amyloid plaque are physiological characteristics of this disease and ?-amyloid(40) fibrillogenesis and disassembly of ?-amyloid(40) fibrils is inhibited by short ?-amyloid congeners containing N-methyl amino acids at alternate residue. N-Methyl-L-serine stimulates hyaluronan production in human skin fibroblasts[2].
N-Methyl-L-serine has been found to stimulate production of high molecular mass hyaluronan in human fibroblasts. This effect was specific to methylated L-serine and not observed with other methylated amino acids. [2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Péter Nemes et al.
Journal of mass spectrometry : JMS, 40(1), 43-49 (2004-12-08)
The association properties of natural and non-natural amino acids were studied in detail using electrospray ionization mass spectrometry. The results show a highly diverse cluster formation behavior of amino acids. There are differences regarding the degree of clustering (average cluster
N-Methyl-L-serine stimulates hyaluronan production in human skin fibroblasts.
Sakai, S., et al.
Skin Pharmacology and Physiology, 12, 276-283 (1999)
D J Gordon et al.
Biochemistry, 40(28), 8237-8245 (2001-07-11)
A potential goal in the prevention or therapy of Alzheimer's disease is to decrease or eliminate neuritic plaques composed of fibrillar beta-amyloid (Abeta). In this paper we describe N-methyl amino acid containing congeners of the hydrophobic "core domain" of Abeta

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service