Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72872

Sigma-Aldrich

Magnesium chloride 0.18 M – HEPES-Na pH 7.5; 0.09M – 2-Propanol 27% (w/v) – Glycerol 10% solution

Synonym(s):

Crystallization solution 12/Kit No 75403

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$230.00

$230.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$230.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

$230.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

pH

7.5±0.2 ( in neat)

density

1.02 g/mL at 20 °C

storage temp.

2-8°C

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service