Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

70797

Sigma-Aldrich

α-Glucosidase from Saccharomyces cerevisiae

lyophilized powder, beige, 4-8 U/mg

Synonym(s):

α-D-Glucosidase, α-D-Glucoside glucohydrolase, Maltase from yeast

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

4-8 U/mg

mol wt

Mr ~63000

color

beige

UniProt accession no.

storage temp.

−20°C

Gene Information

bakers yeast ... MAL12(853209) , MAL32(852602)

Looking for similar products? Visit Product Comparison Guide

General description

only partly soluble in water or buffer

Application

For the determination of α-amylase and the synthesis of various 1′-O-sucrose and 1-O-fructose esters

Biochem/physiol Actions

Hydrolysis of terminal, non-reducing 1→4-linked D-glucose residues with release of D-glucose.

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol p-nitrophenyl-α-D-glucopyranoside per minute at pH 6.8 and 37°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachel Mata et al.
Journal of natural products, 76(3), 468-483 (2013-02-13)
Type II-diabetes mellitus (TII-DM) has been regarded as one of the most important public health problems in all nations in the 21st century. Although allopathic therapies remain the most important for the initial management of TII-DM, herbal remedies have gained
Erik Axelsson et al.
Nature, 495(7441), 360-364 (2013-01-29)
The domestication of dogs was an important episode in the development of human civilization. The precise timing and location of this event is debated and little is known about the genetic changes that accompanied the transformation of ancient wolves into
Shinji Onose et al.
Food chemistry, 138(1), 516-523 (2012-12-26)
1-Deoxynojirimycin (DNJ), a potent α-glycosidase inhibitor, has therapeutic applications in treatments of HIV, Gaucher's disease, and diabetes. DNJ has been extracted from natural sources (mulberry leaves) for therapeutic purposes; however, DNJ ingredients are in limited supply and are costly to
Nicolai Preisler et al.
Molecular genetics and metabolism, 110(3), 287-289 (2013-09-10)
Late-onset Pompe disease is a rare, but potentially treatable metabolic myopathy, and therefore should not be overlooked. However, it is not unusual that patients go undiagnosed for many years. We hypothesized that patients with late-onset Pompe disease may have been
Thomas J Conlon et al.
Human gene therapy. Clinical development, 24(3), 127-133 (2013-09-12)
A biodistribution and toxicology study was performed to test the acute toxicities of intradiaphragmatic injection of a recombinant adeno-associated virus (rAAV) 2/1-human acid alpha-Glucosidase (hGAA) driven by a cytomegalovirus (CMV) promoter (rAAV1-CMV-hGAA) in New Zealand white rabbits and in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service