Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

70543

Sigma-Aldrich

Fluorescent red 610 reactive

suitable for fluorescence, BioReagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H45N4NaO11S
CAS Number:
Molecular Weight:
764.82
MDL number:
UNSPSC Code:
12352200

product line

BioReagent

Assay

≥90.0% (coupling to amines)

solubility

DMF: soluble

fluorescence

λex 610 nm; λem 645 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service