Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69125

Sigma-Aldrich

Resorufin methyl ether

suitable for fluorescence, ≥98.0% (HPLC)

Synonym(s):

7-Methoxy-3H-phenoxazin-3-one, Methoxyresorufin, O7-Methylresorufin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NO3
CAS Number:
Molecular Weight:
227.22
Beilstein:
209529
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (HPLC)

form

crystals

mp

≥220 °C (lit.)

solubility

DMF: soluble
DMSO: soluble
alcohols: soluble

fluorescence

λex 571 nm; λem 585 nm in dealkylase(lit.)

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

COc1ccc2N=C3C=CC(=O)C=C3Oc2c1

InChI

1S/C13H9NO3/c1-16-9-3-5-11-13(7-9)17-12-6-8(15)2-4-10(12)14-11/h2-7H,1H3

InChI key

KNYYMGDYROYBRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to differentiate isozymes of cytochrome P-450

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A Toby A Jenkins et al.
Bioelectrochemistry (Amsterdam, Netherlands), 68(1), 67-71 (2005-06-07)
This paper describes the utilisation of methoxy-resorufin ether as an electrochemical probe for studying cytochrome P450 CYP6G1. Methoxy-resorufin ether is well established as a versatile substrate for cytochrome P450, as its demethylated product, resorufin, is a fluorophore. We show that
D F Staskal et al.
Toxicological sciences : an official journal of the Society of Toxicology, 84(2), 225-231 (2005-01-22)
Dioxins have been shown to bind and induce rodent CYP1A2, producing a dose-dependent hepatic sequestration in vivo. The induction of CYP1A2 activity has been used as a noninvasive biomarker for human exposure to dioxins; while there is a consistent relationship
A G Krainev et al.
Archives of biochemistry and biophysics, 298(1), 198-203 (1992-10-01)
Some eukaryotic cytochromes P450 (P450s) have a series of ionic amino acids, corresponding to Lys250, Arg251, and Lys253 residues in the P450 1A2 sequence. To understand the roles of those ionic amino acids in the catalytic function of P450, three
Xiaowei Zhang et al.
Toxicology and applied pharmacology, 234(3), 306-313 (2008-12-02)
As part of an ongoing effort to understand aryl hydrocarbon receptor (AhR) mediated toxicity in mink, cDNAs encoding for CYP1A1 and the CYP1A2 mixed function monooxygenases were cloned and characterized. In addition, the effects of selected dibenzofurans on the expression
Nicholas E Hadjokas et al.
British journal of pharmacology, 136(3), 347-352 (2002-05-25)
1. Cytochrome P4501A2 (CYP1A2) activates a large number of procarcinogens to carcinogens. Phytochemicals such as flavones can inhibit CYP1A2 activity competitively, and hydroxylated derivatives of flavone (galangin) may be potent, selective inhibitors of CYP1A2 activity relative to CYP1A1 activity. Molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service