Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67678

Sigma-Aldrich

DL-Glyceraldehyde 2-phosphate sodium salt

≥90% (TLC)

Synonym(s):

3-Hydroxy-2-(phosphonooxy)propanal sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7O6P · xNa+
CAS Number:
Molecular Weight:
170.06 (free acid basis)
UNSPSC Code:
12352201
NACRES:
NA.25

Assay

≥90% (TLC)

form

solid

storage temp.

−20°C

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Studies on carbohydrate metabolism in fruit tissues of Citrus acida. I. Detection of the enzymes of the glycolytic cycle in the fruit tissues of Citrus acida.
L J Parekh et al.
Enzymologia, 38(1), 19-22 (1970-01-30)
Phosphorylated sugars. Part 22. Synthesis of 3-deoxy-D-erythro- and 3-deoxy-D-threo-hex-2-ulosonic acid 5-(dihydrogen phosphates).
Trigalo, F., et al.
Journal of the Chemical Society. Perkin Transactions 1, 3, 649-651 (1979)
Regioselective alpha-Phosphorylation of Aldoses in Aqueous Solution Chemistry of alpha-aminonitriles, Part 29. Part 28: Ref. 1, Part 27: Ref. 2. This work was supported by the Skaggs Foundation. S.G. thanks the NASA NSCORT Exobiology program (La Jolla) for a postdoctoral fellowship.
Krishnamurthy et al.
Angewandte Chemie (International ed. in English), 39(13), 2281-2285 (2000-08-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service