Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67516

Sigma-Aldrich

Cacodylate pH 7.0; 0.05 M − Magnesium chloride 18 mM − Spermine 2.25 mM − Cobalt(III)-hexamine 0.9 mM − 2-Methyl-2,4-pentanediol 4.5% solution

Synonym(s):

Crystallization solution 5/Kit-No 80701

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

pH

7.0±0.2 ( in neat)

density

1.01 g/mL at 20 °C

storage temp.

2-8°C

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service