Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

66540193

Sigma-Aldrich

WTSIi071-B

Human iPS Cell Line

Synonym(s):

EBiSC iPSC Line, Human iPSC, Induced Pluripotent Stem Cell Line, iPS Cell, iPSC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509

biological source

human dermis (fibroblast)

Quality Level

reprogramming method

Sendai virus

description

age (45-49)

manufacturer/tradename

EBiSC

gender

female

growth mode

adherent (pluripotent)

technique(s)

cell culture | stem cell: suitable

relevant disease(s)

none

shipped in

dry ice

storage temp.

−196°C

General description

Induced pluripotent stem cells (iPSCs) are adult cells that have been reprogrammed to an embryonic stem cell–like state. The cells can replicate indefinitely or, under controlled conditions, can be differentiated into any other cell type such as nerve, heart or liver cells. Medical researchers are able to use iPS cells to test how different patients might respond to new drugs or to analyse how genetic diseases develop.

The EBiSC stem cell bank is a collection of human iPS cells available to academic and commercial researchers for use in disease modelling and other forms of stem cell research. The initial collection has been generated from a wide range of donors representing specific disease backgrounds and healthy controls. EBiSC has established many routine procedures for collecting, expanding and characterizing human iPS cell lines. The stem cell bank includes iPSC cell lines derived from neurodegenerative diseases (Alzheimer′s Disease, Parkinson′s Disease, Dementia, Motor Neuron Disease (ALS) - and Huntington′s Disease), eye and heart diseases, and lines from healthy control donors for age and sex matching.

Cell Line Origin

Depositor
Wellcome Trust Sanger Institute

Cell Line Description

Derivation
Primary cell type: Fibroblast of dermis
Primary cell developmental stage: Adult
Location of primary tissue procurement: Arm
Reprogramming method
Vector type: Non-integrating
Vector: Sendai virus
Gene list:
KLF4
MYC
POU5F1
SOX2
Is the reprogramming vector detectable: Unknown
Other:From the same donor: WTSIi071-A

Characterization
Analysis of Undifferentiated Cells
Marker expression: POU5F1 (OCT-4)(+)SOX2(+)NANOG (+)
PluriTest: Pluripotency score: 28.62
Novelty score: 1.333
Microarray data: http://www.ebi.ac.uk/arrayexpress/experiments/E-MTAB-4057/

Microbiology / Virus Screening
HIV 1: -
HIV 2: -
Hepatitis B: -
Hepatitis C: -
Mycoplasma: -
Sterility
Inoculation for microbiological growth: No Contaminants Detected
Mycoplasma: Not Detected
Viability: Viable post-cryopreservation

Genotyping
Genotyping
Genome-wide analysis:
Exome sequencing
http://www.ebi.ac.uk/ena/data/view/ERZ122987
STR/Fingerprinting: A 16 allele profile has been recorded and data is available upon request, after cell line purchase.

Linkage

Note: EAUA and CLIP must be completed before order fulfillment

Subculture Routine

Medium: -
Passage method: EDTA
Matrix: Vitronectin
CO2 concentration: 5%
O2 concentration: 20%
Temperature: 37°C

Legal Information

EBiSC is a trademark of Fraunhofer-Gesellschaft

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service