Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

64052

Midas® Slide Carrier (20 Slides)

new

for use with Midas® III-Plus Automated Stainer

Synonym(s):

Hematology Stainer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105202

compatibility

for use with Midas® III-Plus Automated Stainer

shipped in

ambient

General description

Holds 20 slides in MIDAS® Automated Stainer

Application

Holds 20 slides in MIDAS® Automated Stainer
Holds 20 slides, 3 x 1 inch each

Legal Information

MIDAS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service