Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63371

Sigma-Aldrich

2-Oleoyl-1-palmitoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt

≥98.0% (TLC)

Synonym(s):

(9Z)-9-Octadecenoic acid (1R)-1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-2-[(1-oxohexadecyl)oxy]ethyl ester monosodium salt, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) sodium salt, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoglycerol sodium salt, 1-hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′-rac-glycerol) sodium salt, L-α-Phosphatidyl-DL-glycerol, β-oleoyl-γ-palmitoyl sodium salt, PG(16:0/18:1(9Z)), POPG, POPG-Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H76NaO10P
CAS Number:
Molecular Weight:
770.99
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

Assay

≥98.0% (TLC)

form

powder

functional group

ester

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C40H77O10P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-40(44)50-38(36-49-51(45,46)48-34-37(42)33-41)35-47-39(43)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,37-38,41-42H,3-16,19-36H2,1-2H3,(H,45,46);/q;+1/p-1/b18-17-;/t37?,38-;/m1./s1

InChI key

FJXDNGDRHUDFST-XQYKCTAGSA-M

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keiichi Inoue et al.
Nature communications, 10(1), 1993-1993 (2019-05-02)
Microbial rhodopsins are photoreceptive membrane proteins that transport various ions using light energy. While they are widely used in optogenetics to optically control neuronal activity, rhodopsins that function with longer-wavelength light are highly demanded because of their low phototoxicity and
Arin Marchesi et al.
Nature communications, 9(1), 3978-3978 (2018-09-30)
Cyclic nucleotide-gated (CNG) ion channels are non-selective cation channels key to signal transduction. The free energy difference of cyclic-nucleotide (cAMP/cGMP) binding/unbinding is translated into mechanical work to modulate the open/closed probability of the pore, i.e., gating. Despite the recent advances
Yutaro Shiraishi et al.
Nature communications, 9(1), 194-194 (2018-01-18)
The C-terminal region of G-protein-coupled receptors (GPCRs), stimulated by agonist binding, is phosphorylated by GPCR kinases, and the phosphorylated GPCRs bind to arrestin, leading to the cellular responses. To understand the mechanism underlying the formation of the phosphorylated GPCR-arrestin complex
Christopher Aisenbrey et al.
Scientific reports, 10(1), 11652-11652 (2020-07-17)
Magainin 2 and PGLa are cationic, amphipathic antimicrobial peptides which when added as equimolar mixture exhibit a pronounced synergism in both their antibacterial and pore-forming activities. Here we show for the first time that the peptides assemble into defined supramolecular
Muneyuki Matsuo et al.
Scientific reports, 9(1), 6916-6916 (2019-05-08)
DNA is an essential carrier of sequence-based genetic information for all life today. However, the chemical and physical properties of DNA may also affect the structure and dynamics of a vesicle-based model protocell in which it is encapsulated. To test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service