Skip to Content
MilliporeSigma
All Photos(1)

Documents

58199

Sigma-Aldrich

N-Lauroyl-D-sphingosine

≥98.0% (TLC)

Synonym(s):

Cer(d18:1/12:0), (2S,3R,4E)-2-(Lauroylamino)-4-octadecene-1,3-diol, N-Dodecanoyl-D-sphingosine, N-Lauroyl-D-erythro-sphingosine, N-[(1S,2R,3E)-2-Hydroxy-1-(hydroxymethyl)-3-heptadecen-1-yl]dodecanamide, C12 Ceramide (d18:1/12:0), C12-Ceramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H59NO3
CAS Number:
Molecular Weight:
481.79
Beilstein:
1717021
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥98.0% (TLC)

composition

carbon content, 74.79%
hydrogen content, 12.34%
nitrogen content, 2.91%

lipid type

sphingolipids

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C30H59NO3/c1-3-5-7-9-11-13-14-15-16-18-19-21-23-25-29(33)28(27-32)31-30(34)26-24-22-20-17-12-10-8-6-4-2/h23,25,28-29,32-33H,3-22,24,26-27H2,1-2H3,(H,31,34)/b25-23+/t28-,29+/m0/s1

InChI key

HXFPPRPLRSPNIB-VARSQMIESA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Buratta et al.
PloS one, 12(11), e0188840-e0188840 (2017-11-29)
Cells release extracellular vesicles (EVs) in their environment and cellular lipids play an important role in their formation, secretion and uptake. Besides, there is also evidence that EV transferred lipids impact on recipient's cell signaling. Cellular senescence is characterized by
Jin Park et al.
Biochemical pharmacology, 71(8), 1229-1239 (2006-02-02)
Allergic diseases are hypersensitivity disorders that are associated with the generation of specific immunoglobulin E (IgE) in response to environmental allergens. Interleukin (IL)-4, which is primarily produced by the CD4(+) T cells, is an important stimulus for the switching of
Igor O Shmarakov et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1864(5), 629-642 (2019-02-09)
Hepatic stellate cells (HSCs) are non-parenchymal liver cells that characteristically contain multiple retinoid (vitamin A)-containing lipid droplets. In this study, we addressed the metabolic fate of non-retinoid lipids originating from lipid droplet loss during HSCs activation. UPLC/MS/MS and qRT-PCR were
Eunice W M Chin et al.
Neuromolecular medicine, 18(3), 364-377 (2016-07-06)
Rett syndrome (RTT) is a postnatal neurodevelopmental disorder that primarily affects girls. Mutations in the methyl-CpG-binding protein 2 (MECP2) gene account for approximately 95 % of all RTT cases. To model RTT in vitro, we generated induced pluripotent stem cells (iPSCs)
G van Echten-Deckert et al.
Journal of lipid research, 38(12), 2569-2579 (1998-02-11)
De novo synthesis and turnover of endogenous ceramide in cultured skin fibroblasts from patients affected with Farber lipogranulomatosis were studied by biosynthetical labeling of cellular sphingolipids with [14C]serine. The cellular uptake of [14C]serine and incorporation into de novo synthesized ceramide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service