Skip to Content
MilliporeSigma
All Photos(1)

Documents

56946

Sigma-Aldrich

Methyl 3,5-diformyl-1-indolizinecarboxylate

≥90% (HPLC)

Synonym(s):

1-Methoxycarbonyl-3,5-indolizinedicarbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9NO4
CAS Number:
Molecular Weight:
231.20
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Assay

≥90% (HPLC)

form

solid

SMILES string

[H]C(=O)c1cccc2c(cc(C([H])=O)n12)C(=O)OC

InChI

1S/C12H9NO4/c1-17-12(16)10-5-9(7-15)13-8(6-14)3-2-4-11(10)13/h2-7H,1H3

InChI key

ROYXGFIJSNOFGY-UHFFFAOYSA-N

Application

Methyl 3,5-diformyl-1-indolizinecarboxylate is used as a precolumn-labeling reagent(HPLC, HPCE) for amino acids. It is similar to but 3x more sensitive than o-phthaldialdehyde (OPA).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Precolumn-label for amino acids, similar to OPA; but 3 times more sensitive. Described for HPLC and HPCE uses

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caifu Xue et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(5-6), 317-325 (2011-01-21)
In this paper, microdialysis combining with ultra-performance liquid chromatography/quadrupole time-of-flight mass spectrometry (UPLC-Q-TOF/MS) was applied to simultaneously identify components in blood and kidney dialysis after oral administration of Abelmoschus manihot extract. Microdialysis probe was implanted in the jugular vein and
A Pappa-Louisi et al.
Talanta, 93, 279-284 (2012-04-10)
A simple approach for retention modeling of solutes under pH-gradient conditions at various organic contents in the mobile phase is proposed. This approach is based on a retention model arising from the evaluation of the retention data of a set
Marina Patricia Arrieta et al.
Food chemistry, 135(3), 1511-1519 (2012-09-08)
The simultaneous determination of 17 free amino acids and 8 biogenic amines in Alicante Monastrell wines was investigated for the first time. The quantification was carried out by using a RP-HPLC method, based on a pre-column derivatization with o-phthaldialdehyde (OPA)
S. Oguri et al.
Analyst, 120, 63-63 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service