Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56561

SAFC

1-[2-(4-Isothiocyanatophenoxy)ethyl]-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium tosylate

≥95.0% (HPCE), BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H27N3O6S2
CAS Number:
Molecular Weight:
601.69
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

Assay

≥95.0% (HPCE)

fluorescence

λex 407 nm; λem 563 nm in DMF

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.COc2ccc(cc2)-c3cnc(o3)-c4cc[n+](CCOc5ccc(cc5)N=C=S)cc4

InChI

1S/C24H20N3O3S.C7H8O3S/c1-28-21-6-2-18(3-7-21)23-16-25-24(30-23)19-10-12-27(13-11-19)14-15-29-22-8-4-20(5-9-22)26-17-31;1-6-2-4-7(5-3-6)11(8,9)10/h2-13,16H,14-15H2,1H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1

InChI key

AVQZDVMXRPILBE-UHFFFAOYSA-M

General description

Class of highly photostable, water soluble fluorescent labels with large Stoke′s shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient (Ε∼ >20,000 in EtOH)[1].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Litak, P.T.;Kauffman, J.M.
Journal of Heterocyclic Chemistry, 31, 457-457 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service