Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55634

Sigma-Aldrich

7-Hydroxy-4-methyl-3-coumarinylacetic acid

suitable for fluorescence, ≥98.0% (HPLC)

Synonym(s):

4-Methylumbelliferone-3-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10O5
CAS Number:
Molecular Weight:
234.20
Beilstein:
220399
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98.0% (HPLC)

form

solid

solubility

DMF: soluble
DMSO: soluble

fluorescence

λex 360 nm; λem ~450 nm in 0.1 M Tris pH 9.0

suitability

suitable for fluorescence

SMILES string

CC1=C(CC(O)=O)C(=O)Oc2cc(O)ccc12

InChI

1S/C12H10O5/c1-6-8-3-2-7(13)4-10(8)17-12(16)9(6)5-11(14)15/h2-4,13H,5H2,1H3,(H,14,15)

InChI key

OOGKDHCQSZAEQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

suitable as pH-indicator

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Krzysztof Żamojć et al.
Journal of fluorescence, 24(3), 713-718 (2013-12-18)
The fluorescence quenching of different coumarin derivatives (7-hydroxy-4-methylcoumarin, 5,7-dimethoxycoumarin, 7-amino-4-methyl-3-coumarinylacetic acid, 7-ethoxy-4-methylcoumarin, 7-methoxycoumarin, 7-hydroxycoumarin, 7-hydroxy-4-methyl-3-coumarinylacetic acid and 7-amino-4-methylcoumarin) by 4-hydroxy-TEMPO in aqueous solutions at the room temperature was studied with the use of UV-Vis absorption spectroscopy as well as a

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service