Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55345

Sigma-Aldrich

N-Methyl-L-threonine

≥98% (TLC)

Synonym(s):

(2S,3R)-3-Hydroxy-2-(methylamino)butyric acid, N-Me-Thr-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO3
CAS Number:
Molecular Weight:
133.15
Beilstein:
1722279
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥98% (TLC)

form

powder

optical activity

[α]/D -24.0±1.0°, c = 1 in 1 M HCl

composition

carbon, 44.2-46.0%
nitrogen, 10.3-10.8%

color

white

storage temp.

2-8°C

SMILES string

CN[C@@H]([C@@H](C)O)C(O)=O

InChI

1S/C5H11NO3/c1-3(7)4(6-2)5(8)9/h3-4,6-7H,1-2H3,(H,8,9)/t3-,4+/m1/s1

InChI key

CCAIIPMIAFGKSI-DMTCNVIQSA-N

Biochem/physiol Actions

N-Methyl amino acids play an important role in investigations on prevention or therapy of Alzheimer′s disease[1], neurofibrillary tangles or amyloid plaque are physiological characteristics of this disease and ?-amyloid(40) fibrillogenesis and disassembly of ?-amyloid(40) fibrils is inhibited by short ?-amyloid congeners containing N-methyl amino acids at alternate residue.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Gordon et al.
Biochemistry, 40(28), 8237-8245 (2001-07-11)
A potential goal in the prevention or therapy of Alzheimer's disease is to decrease or eliminate neuritic plaques composed of fibrillar beta-amyloid (Abeta). In this paper we describe N-methyl amino acid containing congeners of the hydrophobic "core domain" of Abeta

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service