Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53846

Sigma-Aldrich

1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, 7-nitrobenzofurazan-labeled

BioReagent, suitable for fluorescence, ammonium salt, ≥98.0% (TLC)

Synonym(s):

L-β,γ-Dioleoyl-α-cephaline, 7-nitrobenzofurazan-labeled, N-(7-Nitrobenzofurazan-4-yl)-1,2-dioleoyl-sn-glycero-3-phosphoethanolamine, Phosphatidylethanolamine 18:1, NBD-labeled

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H79N4O11P
CAS Number:
Molecular Weight:
907.12
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

product line

BioReagent

Assay

≥98.0% (TLC)

form

powder

fluorescence

λex 461 nm; λem 529 nm in methanol

suitability

suitable for fluorescence

functional group

ester

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C\CCCCCCCC(=O)OC[C@H](COP(O)(=O)OCCNc1ccc([N+]([O-])=O)c2nonc12)OC(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/C47H79N4O11P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-44(52)58-39-41(61-45(53)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)40-60-63(56,57)59-38-37-48-42-35-36-43(51(54)55)47-46(42)49-62-50-47/h17-20,35-36,41,48H,3-16,21-34,37-40H2,1-2H3,(H,56,57)/b19-17+,20-18+/t41-/m1/s1

InChI key

RKWFXILULDJZBB-JJYVMECISA-N

Application

1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, 7-nitrobenzofurazan-labeled (DOPE,NBD) may be used as a fluorescence label for lipid membranes in studies of membrane structures such as liposomes and lipid rafts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huai-Ying Zhang et al.
Journal of the Royal Society, Interface, 8(54), 127-143 (2010-05-28)
Self-diffusion coefficients of poly(ethylene glycol)2k-derivatized lipids (DSPE-PEG2k-CF) in glass-supported DOPC phospholipid bilayers are ascertained from quantitative fluorescence recovery after photobleaching (FRAP). We developed a first-order reaction-diffusion model to ascertain the bleaching constant, mobile fraction and lipopolymer self-diffusion coefficient D(s) at
André E P Bastos et al.
Chemistry and physics of lipids, 165(5), 577-588 (2012-06-19)
In this work, binary mixtures of phospholipid/ergosterol (erg) were studied using three fluorescent membrane probes. The phospholipid was either saturated (1,2-dipalmitoyl-sn-glycero-3-phosphocholine, DPPC) or monounsaturated (1-palmitoyl-2-dioleoyl-sn-glycero-3-phosphocholine, POPC) phosphatidylcholine, to evaluate the fluorescence properties of the probes in gel, liquid ordered (l(o))
Ligand-based targeted delivery of a peptide modified nanocarrier to endothelial cells in adipose tissue.
Hossen MN, Kajimoto K, Akita H, et al.
J. Controlled Release, 147, 261-268 (2010)
Michela Pisani et al.
The journal of physical chemistry. B, 115(34), 10198-10206 (2011-07-28)
Neutral lipids have received up to now a little attention as genetic material carriers, despite some valuable features, such as the absence of toxicity and the high stability in serum of their complexes with DNA. We have prepared two quaternary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service