Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

53739

Sigma-Aldrich

Laccase from Trametes versicolor

powder, slightly beige, 15 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

form

powder

specific activity

15 U/mg

color

slightly beige

storage temp.

−20°C

InChI

1S/C9H13NO/c1-4-10-7(2)5-9(6-11)8(10)3/h5-6H,4H2,1-3H3

InChI key

NWDZDFOKSUDVJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Laccase catalyzes the oxidation of phenol containing compounds, including lignin, through the reduction of oxygen to water. The presence of mediators will allow the oxidation of non-phenlic compounds as well. The primary function of laccase is to degrade lignin in fungi.

Unit Definition

One unit corresponds to the amount of enzyme which converts 1 μmole of catechol per minute at pH 4.5 and 25 °C

Other Notes

former nomenclature: Coriolus versicolor

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. What is Laccase from Trametes versicolor, Product 53739, soluble in?

    The product is soluble at a concentration of 2 mg/mL in 0.05 M citrate buffer, pH 4.5.

  4. What is the molecular weight of Laccase from Trametes versicolor, Product 53739?

    Unfortunately, neither Sigma-Aldrich nor our supplier has determined the molecular weight of this product.

  5. What is the enzyme content of Laccase from Trametes versicolor, Product 53739?

    According to our supplier, the enzyme content of this Laccase is approximately 10%.

  6. What is the amino acid sequence of Laccase from Trametes versicolor, Product 53739?

    Unfortunately, neither Sigma-Aldrich nor our supplier has determined the amino acid sequence of the enzyme.

  7. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  8. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  9. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Noémie Lalaoui et al.
Chemical communications (Cambridge, England), 49(81), 9281-9283 (2013-09-03)
We report the functionalization of multi-walled carbon nanotube (MWCNT) electrodes by oxidative electropolymerization of pyrrole monomers bearing pyrene and N-hydroxysuccinimide groups. Both polymers were applied to the immobilization and electrical wiring of Trametes versicolor laccase via chemical grafting or non-covalent
Jing Si et al.
Bioresource technology, 128, 49-57 (2012-12-01)
Extracellular laccase (Tplac) from Trametes pubescens was purified to homogeneity by a three-step method, which resulted in a high specific activity of 18.543 Umg(-1), 16.016-fold greater than that of crude enzyme at the same level. Tplac is a monomeric protein
Miharu Seki et al.
Environmental science & technology, 47(1), 390-397 (2012-12-01)
Laccase oxidizes iodide to molecular iodine or hypoiodous acid, both of which are easily incorporated into natural soil organic matter. In this study, iodide sorption and laccase activity in 2 types of Japanese soil were determined under various experimental conditions
Ngoc Han Tran et al.
Bioresource technology, 147, 667-671 (2013-09-17)
Numerous efforts have been made to remove emerging trace organic contaminants, such as pharmaceuticals and personal care products (PPCPs). This study examined the removal of N,N-diethyl-m-toluamide (DEET) by Trametes versicolor laccase and its laccase-mediator systems. Experimental results showed that DEET
Yu-Chan Zhang et al.
Nature biotechnology, 31(9), 848-852 (2013-07-23)
Increasing grain yields is a major focus of crop breeders around the world. Here we report that overexpression of the rice microRNA (miRNA) OsmiR397, which is naturally highly expressed in young panicles and grains, enlarges grain size and promotes panicle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service