Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53517

Sigma-Aldrich

2,10-Dihydroxy-dibenzo[a,j]perylene-8,16-dione

BioReagent, suitable for fluorescence, ≥90% (C)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H14O4
CAS Number:
Molecular Weight:
414.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

Assay

≥90% (C)

suitability

suitable for fluorescence

SMILES string

Oc1ccc-2c(c1)C(=O)c3cccc4c5-c6ccc(O)cc6C(=O)c7cccc(c-2c34)c57

InChI

1S/C28H14O4/c29-13-8-10-16-21(11-13)27(31)19-5-1-3-17-23-15-9-7-14(30)12-22(15)28(32)20-6-2-4-18(26(20)23)24(16)25(17)19/h1-12,29-30H

InChI key

CGRSFQZBJXWSJK-UHFFFAOYSA-N

Application

2,10-Dihydroxy-dibenzo[a,j]perylene-8,16-dione is used as a fluorescence probe to study drug carrier interactions.

Analysis Note

luminescence test: Emmax: 670 nm (Exc.: 620 nm in chloroform)

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Techen et al.
Journal of colloid and interface science, 377(1), 251-261 (2012-04-24)
The well-known cationic surfactant hexadecyltrimethylammonium bromide (CTAB) was used as a model carrier to study drug-carrier interactions with fluorescence probes (5-hexadecanoylaminofluorescein (HAF) and 2,10-bis-(3-aminopropyloxy)dibenzo[a,j]perylene-8,16-dione (NIR 628) by applying ensemble as well as single molecule fluorescence techniques. The impact of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service