Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52944

Sigma-Aldrich

Anti-Rabbit IgG−Abberior® STAR 488 antibody produced in goat

for STED application

Synonym(s):

Abberior® STAR 488-Anti-Rabbit IgG antibody produced in goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rabbit

availability

available only in USA and Canada

concentration

~1 mg/mL

fluorescence

λex 503 nm; λem 524 nm in PBS, pH 7.4

storage temp.

−20°C

General description

Abberior STAR 488 was developed for STED and confocal microscopy in the green spectral region. It is a bright green fluorescent dye. The dye works extremely well with Abberior Instruments Expert Line Microscopes and Leica STED microscopes. It can be very effectively excited with the prominent 488 nm laser line. For STED microscopy, Abberior STAR 488 can be efficiently used with STED Laser wavelengths between 590 - 610 nm. Abberior STAR 488 can substitute dyes like Oregon Green 488, Atto 488 or Alexa Fluor 488. Abberior STAR 488 is the ideal partner for Abberior STAR 440SXP to obtain optimal 2 Color STED results. Best results are obtained with freshly prepared samples.

Photophysical properties (carboxylic acid):
Absorption Maximum, λex [nm]: 503 (PBS pH 7.4), 504 (H2O), 507 (MeOH + 0.1% TFA)
Extinction Coefficient, εmax [M-1cm-1]: 65 000 (PBS pH 7.4), 80 000 (H2O), 85 000 (MeOH + 0.1% TFA)
Correction Factor, CF260 = ε260max: 0,28 (PBS pH 7.4)
Correction Factor, CF280 = ε280max: 0,14 (PBS pH 7.4)
Fluorescence Maximum, λem [nm]: 524 (PBS pH 7.4), 525 (H2O), 531 (MeOH + 0.1% TFA)
Recommended STED Wavelength, λ [nm]: 590 - 610
Fluorescence Quantum Yield, λ: 0,89 (PBS pH 7.4)
Fluorescence Lifetime, τ [ns]: 3,9 (PBS pH 7.4)

Application

Anti-Rabbit IgG-Abberior® STAR 488 antibody has been used as a secondary antibody:
  • for fluorescent immunohistochemistry in prostate cancer cell lines, PPC-1 and TSU-Pr1
  • for immunofluorescence in HeLa and monkey kidney COS-7 cells
  • for STED (stimulated emission depletion) microscopy in OLN-t40-α-syn cells (oligodendroglial cells)

Suitability

Designed and tested for fluorescent super-resolution microscopy

Analysis Note

May contain significant amounts of BSA as stabilizer
unconjugated dye ≤5% of total fluorescence

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
Atto is a trademark of Atto-Tec GmbH
abberior is a registered trademark of Abberior GmbH

Not finding the right product?  

Try our Product Selector Tool.

related product

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Pukaß et al.
Frontiers in cellular neuroscience, 9, 163-163 (2015-05-23)
α-Synuclein (α-syn) positive glial cytoplasmic inclusions (GCI) originating in oligodendrocytes (ODC) are a characteristic hallmark in multiple system atrophy (MSA). Their occurrence may be linked to a failure of the ubiquitin proteasome system (UPS) or the autophagic pathway. For proteasomal
Giuseppe Vicidomini et al.
Methods (San Diego, Calif.), 66(2), 124-130 (2013-07-03)
Stimulation emission depletion (STED) microscopy breaks the spatial resolution limit of conventional light microscopy while retaining its major advantages, such as working under physiological conditions. These properties make STED microscopy a perfect tool for investigating dynamic sub-cellular processes in living
Combination of axitinib and dasatinib for anti-cancer activities in two prostate cancer cell lines.
Peng N
Bangladesh Journal of Pharmacology, 11, 10-10 (2016)
T A Klar et al.
Optics letters, 24(14), 954-956 (2007-12-13)
We overcame the resolution limit of scanning far-field fluorescence microscopy by disabling the fluorescence from the outer part of the focal spot. Whereas a near-UV pulse generates a diffraction-limited distribution of excited molecules, a spatially offset pulse quenches the excited
Tim Grotjohann et al.
Nature, 478(7368), 204-208 (2011-09-13)
Lens-based optical microscopy failed to discern fluorescent features closer than 200 nm for decades, but the recent breaking of the diffraction resolution barrier by sequentially switching the fluorescence capability of adjacent features on and off is making nanoscale imaging routine. Reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service