Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52046

Sigma-Aldrich

Hexaethylene glycol monohexadecyl ether

BioXtra, ≥99.0% (TLC)

Synonym(s):

C16E6, Hexadecylhexaglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15(OCH2CH2)6OH
CAS Number:
Molecular Weight:
506.76
Beilstein:
1806735
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

product line

BioXtra

Assay

≥99.0% (TLC)

form

solid

mol wt

506.76 g/mol

technique(s)

electrophoresis: suitable

mp

39-40 °C

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C28H58O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-17-30-19-21-32-23-25-34-27-28-35-26-24-33-22-20-31-18-16-29/h29H,2-28H2,1H3

InChI key

UOFCAQWHTQFNHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service