Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52043

Sigma-Aldrich

Hexaethylene glycol monodecyl ether

BioXtra, ≥99.0% (GC)

Synonym(s):

C10E6, Decylhexaglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9(OCH2CH2)6OH
CAS Number:
Molecular Weight:
422.60
Beilstein:
1801568
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

product line

BioXtra

Assay

≥99.0% (GC)

mol wt

422.60 g/mol

refractive index

n20/D 1.455

density

0.987 g/mL at 25 °C (lit.)

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C22H46O7/c1-2-3-4-5-6-7-8-9-11-24-13-15-26-17-19-28-21-22-29-20-18-27-16-14-25-12-10-23/h23H,2-22H2,1H3

InChI key

GLGQRQQFWLTGES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Dong et al.
The Journal of biological chemistry, 291(8), 4069-4078 (2015-12-26)
Prostaglandin endoperoxide H synthases (PGHSs), also called cyclooxygenases (COXs), convert arachidonic acid (AA) to PGH2. PGHS-1 and PGHS-2 are conformational heterodimers, each composed of an (Eallo) and a catalytic (Ecat) monomer. Previous studies suggested that the binding to Eallo of
Marvin V Dilworth et al.
Methods (San Diego, Calif.), 147, 3-39 (2018-04-16)
Despite many high-profile successes, recombinant membrane protein production remains a technical challenge; it is still the case that many fewer membrane protein structures have been published than those of soluble proteins. However, progress is being made because empirical methods have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service