Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51554

Sigma-Aldrich

Heptaethylene glycol monododecyl ether

BioXtra, ≥98.0% (TLC)

Synonym(s):

C12E7, Dodecylheptaglycol, Polyoxyethylene (7) lauryl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11(OCH2CH2)7OH
CAS Number:
Molecular Weight:
494.70
Beilstein:
1916066
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

product line

BioXtra

Assay

≥98.0% (TLC)

form

solid

mol wt

494.70 g/mol

technique(s)

enzyme immunoassay: suitable

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C26H54O8/c1-2-3-4-5-6-7-8-9-10-11-13-28-15-17-30-19-21-32-23-25-34-26-24-33-22-20-31-18-16-29-14-12-27/h27H,2-26H2,1H3

InChI key

DWHIUNMOTRUVPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Non-ionic detergent. Can be utilised as a source of C and energy by an organism isolated from activated sludge

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathleen M Clark et al.
Protein science : a publication of the Protein Society, 26(2), 242-257 (2016-10-25)
The function and localization of proteins and peptides containing C-terminal "CaaX" (Cys-aliphatic-aliphatic-anything) sequence motifs are modulated by post-translational attachment of isoprenyl groups to the cysteine sulfhydryl, followed by proteolytic cleavage of the aaX amino acids. The zinc metalloprotease ZMPSTE24 is
Y. Ichikawa et al.
J. Ferment. Technol., 56, 403-403 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service