Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

506-05F

Sigma-Aldrich

Human Lung Fibroblasts: HLF, fetal

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,110.00

$1,110.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
$1,110.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$1,110.00


Please contact Customer Service for Availability

biological source

human lung (normal parenchyma)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 46

morphology

fibroblast

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

asthma; tuberculosis; cardiovascular diseases; cancer

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

HLF provide an excellent model system to study many aspects of human pulmonary function and pathophysiology.

HLF have been utilized in a number of research publications, for example to:
  • Demonstrate differential gene expression patterns between fetal and adult lung fibroblasts, in particular showing that fetal fibroblasts express higher level of TGF-β receptor interacting protein 1 (TRIP-1) and consequently are less able to contract collagen than adult lung fibroblasts, leading to better healing of acute injuries in very immature lungs compared to higher levels of fibrosis observed in older patients (Navarro, 2009), and additionally discover that TRIP-1 regulates lung fibroblast/myofibroblast trans-differentiation (Nyp, 2014)
  • Show that angiotensin II promotes lung fibrosis by activating expression of IL-4, IL-7, and PDGF-D increasing the extracellular matrix formation, and that this process can be blocked by angiotensin receptor inhibitor candesartan (Okada, 2009)
  • Determine that phosphorylation of C/EBPβ contributes to lung fibrosis (Buck, 2011)
  • Demonstrate that increased tissue rigidity activates toll-like receptor 2 and NF-κB in lung fibroblasts and induces expression of cytokines, CXCL1-3, IL-8 and TNF-α (Zheng, 2011)
  • Demonstrate that 2-ethoxyestradiol, a non-estrogenic estradiol metabolite analog, inhibits cell growth and retards the progression of pulmonary fibrosis and hypertension (Tofovic, 2008, 2009)
  • Identify the hypoxia-induced ROS/RhoA–ROCK dependent actin reorganization as a mediator of myofibroblast transformation and collagen synthesis in lung fibrosis (Ni, 2013)

Cell Line Origin

Lungs

Application

pulmonary function, gene expression, healing and repair process, differentiation, extracellular matrix production, growth factor and cytokine activation, actin reorganization, synthesis of type II collagen, elastin, and proteoglycans, airway remodeling, pharmaceutical development

Components

Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 1st passage, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 12 doublings

Subculture Routine

Please refer to the HLF Culture Protocol.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Store the cryovials in a liquid nitrogen storage tank immediately upon arrival.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service