Skip to Content
MilliporeSigma
All Photos(1)

Documents

50552

Sigma-Aldrich

Calcium phosphate

Reagent for transient & stable DNA transfections

Synonym(s):

β-Calcium phosphate tribasic, β-Tricalcium phosphate, tert-Calcium phosphate, tri-Calcium (ortho)phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ca3O8P2
CAS Number:
Molecular Weight:
310.18
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for molecular biology

form

powder

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

room temp

SMILES string

[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/3Ca.2H3O4P/c;;;2*1-5(2,3)4/h;;;2*(H3,1,2,3,4)/q3*+2;;/p-6

InChI key

QORWJWZARLRLPR-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

General description

Calcium phosphate is an amorphous, tasteless powder containing excess of calcium oxide. It is commonly used to introduce DNA and transfect eukaryotic cells for molecular biology experiments.

Application

Suitable for
  • transient and stable transfection of wide range of cell types
  • drug delivery systems

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael D Weiden et al.
PloS one, 9(10), e108476-e108476 (2014-10-02)
While exploring the effects of aerosol IFN-γ treatment in HIV-1/tuberculosis co-infected patients, we observed A to G mutations in HIV-1 envelope sequences derived from bronchoalveolar lavage (BAL) of aerosol IFN-γ-treated patients and induction of adenosine deaminase acting on RNA 1
S I Roohani-Esfahani et al.
Acta biomaterialia, 8(11), 4162-4172 (2012-07-31)
This is the first reported study to prepare highly porous baghdadite (Ca₃ZrSi₂O₉) scaffolds with and without surface modification and investigate their ability to repair critical-sized bone defects in a rabbit radius under normal load. The modification was carried out to
A Butscher et al.
Acta biomaterialia, 9(2), 5369-5378 (2012-10-17)
Powder based three-dimensional printing (3DP) allows great versatility in material and geometry. These characteristics make 3DP an interesting method for the production of tissue engineering scaffolds. However, 3DP has major limitations, such as limited resolution and accuracy, hence preventing the
Françoise Borcard et al.
Journal of medicinal chemistry, 55(18), 7988-7997 (2012-08-18)
To control the selective adhesion of human endothelial cells and human serum proteins to bioceramics of different compositions, a multifunctional ligand containing a cyclic arginine-glycine-aspartate (RGD) peptide, a tetraethylene glycol spacer, and a gallate moiety was designed, synthesized, and characterized.
Muwan Chen et al.
International journal of nanomedicine, 7, 4285-4297 (2012-08-21)
Bone tissue engineering implants with sustained local drug delivery provide an opportunity for better postoperative care for bone tumor patients because these implants offer sustained drug release at the tumor site and reduce systemic side effects. A rapid prototyped macroporous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service