Fluram (fluorescamine) is a non-fluorescent reagent that reacts readily under mild conditions with primary amines in amino acids and peptides and other molecules to form stable, highly fluorescent compounds. Fluorescamine is useful for the fluorometric assay of amino acids, protein, and proteolytic enzymes and as a pre-column derivatization reagent. It effectively blocks newly generated amino termini in protein sequence analyses.
Non-fluorescent reagent that reacts readily under mild conditions with primary amines in amino acids and peptides to form stable, highly fluorescent compounds. Low background due to hydrolysis. Useful for the fluorometric assay of amino acids, protein, and proteolytic enzymes. Effectively blocks newly generated amino termini in protein sequence analyses.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Aging is accompanied by the accumulation of oxidized proteins. To remove them, cells employ the proteasomal and autophagy-lysosomal systems; however, if the clearance rate is inferior to its formation, protein aggregates form as a hallmark of proteostasis loss. In cells
Here we report a fast and effective method to visualize interactive proteins across intact mammalian cells via on-site formation of fluorescence using instant reaction of non-fluorescent fluorescamine with primary amines on proteins. Without interference by fluorescence background, this fluorogenic labelling
The Urey organic and oxidant detector consists of a suite of instruments designed to search for several classes of organic molecules in the martian regolith and ascertain whether these compounds were produced by biotic or abiotic processes using chirality measurements.
Journal of agricultural and food chemistry, 56(22), 10811-10816 (2008-11-06)
Photoinduced decarboxylation via homolytic cleavage of the ester linkage generating two benzyl radicals being recoupled is known to be a major photolytic pathway of the insecticide fenvalerate in aqueous or organic solvents. A highly sensitive and selective fluorescence spectroscopic method
This account highlights some of our recent activities focused on developing microfluidic technologies for application in high-throughput and high-information content chemical and biological analysis. Specifically, we discuss the use of continuous and segmented flow microfluidics for artificial membrane formation, the