Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45260

Sigma-Aldrich

Eosin B

for microscopy (Fl., Hist.), adsorption and fluorescent indicator

Synonym(s):

4′,5′-Dibromo-2′,7′-dinitrofluorescein disodium salt, Acid Red 91, Eosin Scarlet, Eosin bluish, Saffrosine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H6N2O9Br2Na2
CAS Number:
Molecular Weight:
624.06
Colour Index Number:
45400
Beilstein:
4121964
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

adsorption and fluorescent indicator
for microscopy (Fl., Hist.)

technique(s)

titration: suitable

λmax

509-524 nm

SMILES string

[Na+].[Na+].[O-]C(=O)c1ccccc1C2=C3C=C(C(=O)C(Br)=C3Oc4c(Br)c([O-])c(cc24)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C20H8Br2N2O9.2Na/c21-14-16(25)11(23(29)30)5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(24(31)32)17(26)15(22)19(10)33-18(9)14;;/h1-6,25H,(H,27,28);;/q;2*+1/p-2

InChI key

GYYTYUGGVBYJHE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Eosin B is a fluorescent red dye that is used in microscopy to visualise cytoplasm and proteins like collagen.

Application

Eosin B is an adsorption and fluorescent indicator in microscopy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Wang et al.
Electrophoresis, 31(23-24), 3808-3815 (2010-11-17)
In this study, we describe an effective visualizing technique for proteins in SDS-PAGE based on the organic dye, Eosin B, the sensitivity of which can be further strengthened by the addition of magnesium to the staining solution after electrophoresis. The
A A Waheed et al.
Analytical biochemistry, 287(1), 73-79 (2000-11-18)
Eosin B and eosin Y have been used to estimate micro- and submicrogram quantities of proteins respectively as shown in our previous reports. In the present study we describe the mechanism of eosin binding to proteins. At pH lower than
Chloé E Atreya et al.
The Journal of biological chemistry, 278(16), 14092-14100 (2003-01-31)
Protozoal parasites are unusual in that their thymidylate synthase (TS) and dihydrofolate reductase (DHFR) enzymes exist on a single polypeptide. In an effort to probe the possibility of substrate channeling between the TS and DHFR active sites and to identify
Jishu Naskar et al.
Organic & biomolecular chemistry, 9(19), 6610-6615 (2011-08-13)
Self-assembling short peptides can offer an opportunity to make useful nano-/microstructures that find potential application in drug delivery. We report here the formation of multivesicular structures from self-assembling water-soluble synthetic amphiphilic dipeptides containing a glutamic acid residue at the C-terminus.
H J Shin et al.
Biochimica et biophysica acta, 1481(1), 139-146 (2000-09-27)
Among various dyes including congo red, thioflavin S, thioflavin T, eosin, rhodamine 6G, and phenol red, the eosin was the only dye that induced self-oligomerization of alpha-synuclein in the presence of a chemical coupling reagent of N-(ethoxycarbonyl)-2-ethoxy-1, 2-dihydroquinoline. To analyze

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service