Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4465863

Sigma-Aldrich

CYP450 Extended Panel - 100 Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.24

technique(s)

LC/MS: suitable

storage temp.

−20°C

General description

The SCIEX iChemistry® CYP450 Peptide Standards Extended Panel Human Induction 100 Assay Kit provides a complete protocol, instrument data acquisition methods, processing methods and stable isotope-labeled synthetic peptide standards for the analysis of seven human CYP450 isoforms (1A2, 2B6, 2C9, 2C19, 2E1, 3A4, 3A5) for induction studies using LC/MS/MS.  Kit contains enough reagents for 100 assays.  Trypsin TPCK (500 μg) and Peptide C18 Column must be ordered separately.

Analysis Note

To view the Protocol for the CYP450 Protein Assay - Human Induction Kit Extended Panel, please visit this Protocol link.

A Technical Note on the use of the CYP450 Protein Assay - Human Induction Kit Extended Panel is available at this Tech Note link.

Legal Information

iChemistry is a registered trademark of AB Sciex Pte. Ltd.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

68.0 °F

Flash Point(C)

20 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service