Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4390733

Sigma-Aldrich

iTRAQ® Reagent - 8PLEX 25U Bulk Pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.24

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Includes 8 tubes. Each tube contains 25 units of an individual 8-plex reagent. This kit contains the reagents in the equivalent Sciex iChemistry® product number 4393528.

Analysis Note

To view the Protocol for the iTRAQ Reagents 8-plex Kit, please visit this Protocol link.

To view the Chemistry Quick Reference Card for the iTRAQ Reagents, please visit this Chemistry Quick Reference Card link.

Legal Information

iChemistry is a registered trademark of AB Sciex Pte. Ltd.
iTraq is a registered trademark of AB Sciex Pte. Ltd.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service