Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

433R-1

Sigma-Aldrich

IMP3 (EP286) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$246.00
0.1 ML
$270.00
0.5 ML
$619.00
7 ML
$706.00
1 ML
$935.00

$246.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$246.00
0.1 ML
$270.00
0.5 ML
$619.00
7 ML
$706.00
1 ML
$935.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$246.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP286, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (433R-14)
vial of 0.5 mL concentrate (433R-15)
bottle of 1.0 mL predilute (433R-17)
vial of 1.0 mL concentrate (433R-16)
bottle of 7.0 mL predilute (433R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG

control

pancreatic ductal adenocarcinoma, placenta

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, membranous

Gene Information

human ... IMP3(55272)

General description

Insulin-like growth factor II mRNA binding protein 3 (IMP3) is an oncofetal RNA-binding protein that regulates targets such as insulin-like growth factor-2 and beta-actin. IMP3 expression is at its highest during embryogenesis and silenced in normal human tissues (fibroblasts, lymphocytes, and testes are exceptions).1 IMP3 plays a major role in early embryogenesis involving the development of the intestine, thymus, pancreas, and kidneys.2 IMP3 mRNA transcript and protein have been demonstrated in high levels in pancreatic cancer tissues but not in benign lesions of the pancreas, chronic pancreatitis, or normal pancreatic tissues.1 In a study of IMP3 being used to distinguish between pancreatic ductal adenocarcinoma from chronic sclerosing pancreatitis, IMP3 was found to have high sensitivity and specificity for pancreatic ductal adenocarcinoma and detected pancreatic ductal adenocarcinoma metastases.2 IMP3 expression has also been found in a variety of other human cancers including renal cell carcinoma, adenocarcinoma of the uterine cervix, endometrial carcinoma, adenocarcinoma of the esophagus, malignant melanoma, Merkel cell carcinoma, urothelial carcinoma, neuroendocrine carcinoma of the lung, adenocarcinoma of the pancreas, and triple negative breast cancer.1,2

Quality


IVD

IVD

IVD

RUO

Linkage

IMP3 Positive Control Slides, Product No. 433S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service