Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

431R-1

Sigma-Aldrich

Islet-1 (EP283) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP283, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (431R-17)
bottle of 7.0 mL predilute (431R-18)
vial of 0.1 mL concentrate (431R-14)
vial of 0.5 mL concentrate (431R-15)
vial of 1.0 mL concentrate (431R-16)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... ISL1(3670)

General description

Islet-1, a human insulin gene enhancer-binding protein, is a transcription factor involved in the differentiation of the neuroendocrine pancreatic cells.1-3 Islet-1 plays an important role in the embryogenesis and differentiation of the insulin producing pancreatic beta cells within the islets of Langerhans.3 Neuroendocrine tumors can arise from a variety of primary sites, most commonly in the bronchopulmonary system, gastrointestinal (GI) tract, and pancreas.2 Neuroendocrine tumors of the GI tract and pancreas tend to be slow growing and indolent, and may metastasize to the liver before it is detected.2 Islet-1 exhibits strong nuclear staining in the islets of normal pancreas and tumor cells of the pancreatic neuroendocrine tumor.1-3 Islet-1 has been shown to be a reliable marker for the detection of primary and metastatic pancreatic neuroendocrine neoplasms.1-3

Quality


IVD

IVD

IVD

RUO

Linkage

Islet-1 Positive Control Slides, Product No. 431S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abbas Agaimy et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 26(7), 995-1003 (2013-03-19)
The human insulin gene enhancer-binding protein islet-1 (ISL1) is a transcription factor involved in the differentiation of the neuroendocrine pancreatic cells. Recent studies identified ISL1 as a marker for pancreatic well-differentiated neuroendocrine neoplasms. However, little is known about ISL1 expression
Anja M Schmitt et al.
The American journal of surgical pathology, 32(3), 420-425 (2008-02-28)
Tracing the origin of a metastasis of a neuroendocrine carcinoma is a challenge. The transcription factors Cdx2 and TTF1 have been found to be helpful in identifying well-differentiated neuroendocrine tumors of gastrointestinal and pulmonary origin, respectively. So far, such a
Jamie Koo et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 25(6), 893-901 (2012-03-06)
Neuroendocrine tumors can present as liver metastases before discovery of the primary tumor. Islet 1 and PAX8 have recently been proposed as markers for neuroendocrine tumors of pancreatic origin. In this study, we compared the utility of Islet 1 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service